Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

701726

Sigma-Aldrich

Chlorzoxazone-2-13C-hydroxy-18O

99 atom % 13C, 95 atom % 18O, 97% (CP)

Synonym(s):

5-Chloro-2-hydroxybenzoxazole-2-13C,18O(hydroxy-18O)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C613CH4ClNO18O
Molecular Weight:
172.56
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
95 atom % 18O

assay

97% (CP)

form

powder

mp

191-192  °C

mass shift

M+3

SMILES string

[18OH][13c]1nc2cc(Cl)ccc2o1

InChI

1S/C7H4ClNO2/c8-4-1-2-6-5(3-4)9-7(10)11-6/h1-3H,(H,9,10)/i7+1,10+2

InChI key

TZFWDZFKRBELIQ-NJBKSYGFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service