Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

701351

Sigma-Aldrich

1-(Methylamino)-3-phenyl-2-propyne

technical grade

Synonym(s):

N-Methyl-3-phenyl-2-propyn-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

form

liquid

refractive index

n20/D 1.578

density

0.991 g/mL at 25 °C

SMILES string

CNCC#Cc1ccccc1

InChI

1S/C10H11N/c1-11-9-5-8-10-6-3-2-4-7-10/h2-4,6-7,11H,9H2,1H3

InChI key

HXHNCCXRHHKYJM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

219.9 °F

flash_point_c

104.4 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service