Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

700401

Sigma-Aldrich

ChiroSolv® Resolving Kit, Complete Base Series, for solid racemates

Synonym(s):

ChiroSolve Resolving Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352000

Application

ChiroSolve Kits are high throughput disposable kits that allow rapid screening to find the optimum resolution method for a given racemate. Each kit offers 96 unique combinations of chiral resolving agents and solvents to screen against. There are 4 kits in the Acid Series and 4 kits in the Base series, allowing for a maximum of 768 combinations. Results can be achieved within 24 hours. Some of the solutes included in this kit are shown below. For a complete list of components, please contact your local Sigma-Aldrich Technical Service office. See Application Guide or More Information links for information on the entire kit range.
ChiroSolve Kits are high throughput disposable kits that allow rapid screening to find the optimum resolution method for a given racemate. Each kit offers 96 unique combinations of chiral resolving agents and solvents to screen against. There are 4 kits in the Acid Series and 4 kits in the Base series, allowing for a maximum of 768 combinations. The kits are available in two different formats in each series, one for solid racemates, the other for liquid racemates. Results can be achieved within 24 hours. Some of the solutes included in this kit are shown below. For a complete list of components, or Application Guide, please contact your local Sigma-Aldrich Technical Service office.

Legal Information

ChiroSolv is a registered trademark of Vaidya, Niteen A

Kit Components Also Available Separately

Product No.
Description
SDS

  • ChiroSolv® Resolving Kit, Base Series 1, for solid racemates

  • ChiroSolv® Resolving Kit, Base Series 2, for solid racemates

  • ChiroSolv® Resolving Kit, Base Series 3, for solid racemates

  • ChiroSolv® Resolving Kit, Base Series 4, for solid racemates


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service