Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699543

Sigma-Aldrich

Acetonitrile-d3

"Special HOH", ≥99.8 atom % D

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein/REAXYS Number:
1740230
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

≥99.8 atom % D

assay

99% (CP)

form

liquid

expl. lim.

3.0-16 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.02% water
water

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

mp

-46 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetonitrile-d3 may be used as:
  • IR and NMR solvent.[1]
  • Probe for zeolite acidity.[2]
  • Cartridge eluent in HPLC– SPE (solid-phase extraction)–NMR technique.[3]
These solvents have an HOD peak which is less than 1% of the HOH peak to minimize potential exchange with an analyte. "Special HOH" also meet our standard water specifications for NMR solvents.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maja Lambert et al.
Magnetic resonance in chemistry : MRC, 43(9), 771-775 (2005-07-29)
The HPLC-SPE-NMR technique was used for the analysis of a root-bark extract of Croton membranaceus. The components of the extract were separated on an analytical-size reversed-phase HPLC column, the chromatographic peaks were trapped on SPE (solid-phase extraction) cartridges after post-column
Acetonitrile-d3 as a probe of Lewis and Broensted acidity of zeolites.
Pelmenschikov AG, et al.
The Journal of Physical Chemistry, 97(42), 11071-11074 (1993)
Organic deuterium compounds: XVI. Synthesis of α-deuterated alkyl nitriles.
Leitch LC.
Canadian Journal of Chemistry, 35(4), 345-347 (1957)
Proton polarizability caused by collective proton motion in intramolecular chains formed by two and three hydrogen bonds. Implications for charge conduction in bacteriorhodopsin.
Brzezinski B, et al.
The Journal of Physical Chemistry, 91(11), 3077-3080 (1987)
Dynamics of a wetting liquid in nanopores: An optical Kerr effect study of the dynamics of acetonitrile confined in sol-gel glasses.
Loughnane BJ, et al.
J. Chem. Phys. , 111(11), 5116-5123 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service