Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699306

Sigma-Aldrich

Pentacene-N-sulfinyl-tert-butylcarbamate

99% (HPLC)

Synonym(s):

13,6-(Epithioimino)pentacene-16-carboxylic acid, 6,13-dihydro-, tert butyl ester, 15-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H23NO3S
CAS Number:
Molecular Weight:
441.54
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99% (HPLC)

form

solid

mp

>300 °C

solubility

THF: soluble 1 mg/mL
ethanol: slightly soluble 1 mg/mL
isopropanol: slightly soluble 1 mg/mL
methanol: slightly soluble 1 mg/mL

SMILES string

CC(C)(C)OC(=O)N1C2c3cc4ccccc4cc3C(c5cc6ccccc6cc25)S1=O

InChI

1S/C27H23NO3S/c1-27(2,3)31-26(29)28-24-20-12-16-8-4-6-10-18(16)14-22(20)25(32(28)30)23-15-19-11-7-5-9-17(19)13-21(23)24/h4-15,24-25H,1-3H3

InChI key

VQUHUWBRYQBGLV-UHFFFAOYSA-N

Application

Photopatternable pentacene precursor highly solube in apolar organic solvents (halogenated solvents, THF). Converted to pentacene in thin (e.g. spin-cast) films by heating under N2 atmosphere for one hour at 150 °C. Spin-coated thin films of the pentacene precursor can be patterned by exposure to UV light in the presence of a photoacid generator (e.g. 531014), followed by a brief heating (5 min. at 130 °C and washing away of an un-converted precursor. Organic thin film transistors fabricated by solution processing of this precursor showed mobilities of 0.13 cm2V-1s-1 and on/off ratio 2 x 105.[1]

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin P Weidkamp et al.
Journal of the American Chemical Society, 126(40), 12740-12741 (2004-10-08)
We demonstrate that modifying pentacene to incorporate an acid-labile moiety into its molecular structure leads to a new precursor that can be easily deposited, photopatterned, and processed via wet-chemical methods to produce organic semiconducting devices exhibiting good electrical characteristics. Acidic

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service