Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

699039

Sigma-Aldrich

Deuterium bromide solution

30-35 wt. % in acetic acid-d4, 98 atom % D

Synonym(s):

DBr solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DBr
Molecular Weight:
81.92
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

concentration

30-35 wt. % in acetic acid-d4

mass shift

M+1

SMILES string

Br[2H]

InChI

1S/BrH/h1H/i/hD

InChI key

CPELXLSAUQHCOX-DYCDLGHISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

176.0 °F - closed cup

flash_point_c

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service