698474
Bis(4-methoxyphenyl)chlorophosphine
95%
Synonym(s):
P,P-Bis(4-methoxyphenyl)phosphinous chloride
About This Item
Recommended Products
assay
95%
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
62-66 °C
functional group
phosphine
SMILES string
COc1ccc(cc1)P(Cl)c2ccc(OC)cc2
InChI
1S/C14H14ClO2P/c1-16-11-3-7-13(8-4-11)18(15)14-9-5-12(17-2)6-10-14/h3-10H,1-2H3
InChI key
YTFQUBRFOJIJOZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Allylic substitution
- Intermolecular Pauson-Khand reaction of symmetrically substituted alkynes
- Hydrogenation
- Intramolecular α-arylation of aldehydes
Reactant for:
- Alcoholysis of phosphane-boranes
- Probing hemilabile sites of Bis(phosphine) monoxide ligands
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service