698393
1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Synonym(s):
(R,R)-Et-DUPHOS-Rh
About This Item
Recommended Products
form
powder
functional group
fluoro
phosphine
triflate
SMILES string
[Rh+].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.CC[C@@H]2CC[C@@H](CC)P2c3ccccc3P4[C@H](CC)CC[C@H]4CC
InChI
1S/C22H36P2.C8H12.CHF3O3S.Rh/c1-5-17-13-14-18(6-2)23(17)21-11-9-10-12-22(21)24-19(7-3)15-16-20(24)8-4;1-2-4-6-8-7-5-3-1;2-1(3,4)8(5,6)7;/h9-12,17-20H,5-8,13-16H2,1-4H3;1-2,7-8H,3-6H2;(H,5,6,7);/q;;;+1/p-1/b;2-1-,8-7-;;/t17-,18-,19-,20-;;;/m1.../s1
InChI key
HZLILTNLWVOBFS-KYOOHHHUSA-M
Application
Catalyst for:
- Stereoselective synthesis of δ-amino acid derivatives via asymmetric hydrogenation of acetylaminopentenoic acid derivatives
- Stereoselective synthesis of manzacidins A and C via stereoselective hydrogenation
- Stereoselective synthesis of tetracyclic core of manzamine A via Rh-catalyzed asymmetric hydrogenation, diastereoselective Diels-Alder reaction, Eschenmoser-Tanabe fragmentation, Chang′s amide formation, and Hofmann rearrangement
- Asymmetric preparation of chiral Cbz-aminodifluorobutyric acid Me ester and its analogs
- Biphasic catalytic hydrogenations in ionic liquids with addition of water as a second solvent
- Preparation of cyclobutane-containing amino acids via asymmetric hydrogenations of cyclobutyl enamides
- Asymmetric preparation of both enantiomers of (dimethoxycoumaryl)alanine as suitable fluorescent peptide labels
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service