Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698318

Sigma-Aldrich

Diethyl-2,2′-13C2-amine-15N

99 atom % 13C, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(13CH3CH2)215NH
Molecular Weight:
76.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C
98 atom % 15N

assay

99% (CP)

bp

55 °C (lit.)

mp

-50 °C (lit.)

density

0.735 g/mL at 25 °C

mass shift

M+3

SMILES string

[13CH3]C[15NH]C[13CH3]

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1+1,2+1,5+1

InChI key

HPNMFZURTQLUMO-NLQHJZJRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-9.4 °F - closed cup

flash_point_c

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service