Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698296

Sigma-Aldrich

1,2-Bis(diethylphosphino)ethane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24P2
CAS Number:
Molecular Weight:
206.25
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

reaction suitability

core: iron
reagent type: catalyst

density

0.880 g/mL at 25 °C

SMILES string

CCP(CC)CCP(CC)CC

InChI

1S/C10H24P2/c1-5-11(6-2)9-10-12(7-3)8-4/h5-10H2,1-4H3

InChI key

MIOCUERTSIJEDP-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

147.0 °F

flash_point_c

63.9 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service