Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69784

Sigma-Aldrich

L-Lactic acid tetrabutylammonium salt solution

~70% in H2O (NT)

Synonym(s):

Tetrabutylammonium L-lactate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H41NO3
Molecular Weight:
331.53
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

liquid

optical activity

[α]20/D −4.5±0.5°, neat

concentration

~70% in H2O (NT)

impurities

~30% water

refractive index

n20/D 1.438

SMILES string

C[C@H](O)C([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C3H6O3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2(4)3(5)6/h5-16H2,1-4H3;2,4H,1H3,(H,5,6)/q+1;/p-1/t;2-/m.0/s1

InChI key

MSLTZKLJPHUCPU-WNQIDUERSA-M

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service