Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

697621

Sigma-Aldrich

[(Triisopropylsilyl)acetylene]sulfur pentafluoride

95%

Synonym(s):

Pentafluoro[(triisopropylsilyl)ethynyl]sulfur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21F5SSi
CAS Number:
Molecular Weight:
308.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.414

density

1.076 g/mL at 25 °C

SMILES string

CC(C)[Si](C#CS(F)(F)(F)(F)F)(C(C)C)C(C)C

InChI

1S/C11H21F5SSi/c1-9(2)18(10(3)4,11(5)6)8-7-17(12,13,14,15)16/h9-11H,1-6H3

InChI key

OHFCQWVMVRPEHF-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service