Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696994

Sigma-Aldrich

2,3-Dimethyl-2,3-dinitrobutane-d12

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2C(NO2)C(CD3)2NO2
Molecular Weight:
188.24
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

assay

97% (CP)

mp

214-215 °C (dec) (lit.)

mass shift

M+12

SMILES string

[2H]C([2H])([2H])C([N+]([O-])=O)(C([2H])([2H])[2H])C([N+]([O-])=O)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C6H12N2O4/c1-5(2,7(9)10)6(3,4)8(11)12/h1-4H3/i1D3,2D3,3D3,4D3

InChI key

DWCLXOREGBLXTD-MGKWXGLJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service