Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696986

Sigma-Aldrich

Potassium carbonate-18O3

50 atom % 18O, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2C18O3
Molecular Weight:
144.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

50 atom % 18O

assay

97% (CP)

mol wt

141.21 by amino acid sequence (50 atom % 18O)

mp

891 °C (lit.)

mass shift

M+6

SMILES string

[K+].[K+].[18O-]C([18O-])=[18O]

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2/i2+2,3+2,4+2;;

Inchi Key

BWHMMNNQKKPAPP-JYIUEGOPSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service