Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696978

Sigma-Aldrich

1,2-Dicyano-13C2-benzene

99 atom % 13C, 98% (CP)

Synonym(s):

Phthalonitrile-dicyano-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(13CN)2
Molecular Weight:
130.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

98% (CP)

mp

137-139 °C (lit.)

mass shift

M+2

SMILES string

N#[13C]c1ccccc1[13C]#N

InChI

1S/C8H4N2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H/i5+1,6+1

InChI key

XQZYPMVTSDWCCE-MPOCSFTDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

323.6 °F - closed cup

flash_point_c

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service