Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696900

Sigma-Aldrich

Water solution

0.005N in sulfuric acid, suitable for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages

grade

HPLC grade

form

liquid

concentration

0.005N in sulfuric acid

technique(s)

HPLC: suitable

density

1.000 g/mL at 25 °C

application(s)

food and beverages

SMILES string

O

InChI

1S/H2O/h1H2

Inchi Key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is water containing 0.005N sulfuric acid that can be employed for HPLC.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service