Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

696854

Sigma-Aldrich

Tripotassium 5,5′,5′′-[2,2′,2′′-nitrilotris(methylene)tris(1H-benzimidazole-2,1-diyl)]tripentanoate hydrate

95%

Synonym(s):

(BimC4A)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H42K3N7O6 · xH2O
Molecular Weight:
822.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

174-180 °C

SMILES string

O.[K+].[K+].[K+].[O-]C(=O)CCCCn1c(CN(Cc2nc3ccccc3n2CCCCC([O-])=O)Cc4nc5ccccc5n4CCCCC([O-])=O)nc6ccccc16

InChI

1S/C39H45N7O6.3K.H2O/c47-37(48)19-7-10-22-44-31-16-4-1-13-28(31)40-34(44)25-43(26-35-41-29-14-2-5-17-32(29)45(35)23-11-8-20-38(49)50)27-36-42-30-15-3-6-18-33(30)46(36)24-12-9-21-39(51)52;;;;/h1-6,13-18H,7-12,19-27H2,(H,47,48)(H,49,50)(H,51,52);;;;1H2/q;3*+1;/p-3

InChI key

QRRSLHKLVBVPSD-UHFFFAOYSA-K

Application

Water soluble benzimidazole ligand for superior enhancement of copper(I) catalyzed azide-alkyne cycloaddition (CuAAC) reactions[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentin O Rodionov et al.
Journal of the American Chemical Society, 129(42), 12696-12704 (2007-10-05)
Tris(2-benzimidazolylmethyl)amines have been found to be superior accelerating ligands for the copper(I)-catalyzed azide-alkyne cycloaddition reaction. Candidates bearing different benzimidazole N-substituents as well as benzothiazole and pyridyl ligand arms were evaluated by absolute rate measurements under relatively dilute conditions by aliquot

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service