Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

696838

Sigma-Aldrich

7-Azaindole N-oxide hemihydrate

97%

Synonym(s):

1H-Pyrrolo[2,3-b]pyridine, 7-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2O · 0.5H2O
CAS Number:
Molecular Weight:
143.14
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

102-118 °C

SMILES string

O.[O-][n+]1cccc2cc[nH]c12.[O-][n+]3cccc4cc[nH]c34

InChI

1S/2C7H6N2O.H2O/c2*10-9-5-1-2-6-3-4-8-7(6)9;/h2*1-5,8H;1H2

InChI key

CNYNVSPJOUGIJA-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service