Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

696064

Sigma-Aldrich

(S)-C3-TunePhos

Synonym(s):

(S)-Bis(diphenylphosphino)-7,8-dihydro-6H-dibenzo[f,h][1,5]dioxonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H32O2P2
CAS Number:
Molecular Weight:
594.62
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

powder

InChI

1S/C39H32O2P2/c1-5-16-30(17-6-1)42(31-18-7-2-8-19-31)36-26-13-24-34-38(36)39-35(41-29-15-28-40-34)25-14-27-37(39)43(32-20-9-3-10-21-32)33-22-11-4-12-23-33/h1-14,16-27H,15,28-29H2

InChI key

GTIXSUJKFAATAE-UHFFFAOYSA-N

Application

Chiral Quest Phosphine Ligands for Asymmetric Hydrogenation

Reactant for:
  • Asymmetric hydrogenation of ketones with TunePhos/diamine/ruthenium complexes
  • Stereoselective preparation of arylalkyl esters via enantioselective hydrogenation of enol acetates

Precatalyst for:
  • Enantioselective iridium-catalyzed carbonyl allylation from alcohol or aldehyde by transfer hydrogenative coupling of allyl acetate

Catalyst for:
  • Enantioselective hydrogenation of a-keto esters to a-hydroxy esters using Ru-tunephos catalysts
Provides comparable or superior enantioselectivities and catalytic abilities to BINAP in Ru-catalyzed asymmetric hydrogenations of β-keto esters[1], cyclic β-(acylamino)acrylates[2] and a-phthalimide ketones[3].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of chiral bisphosphines with tunable bite angles and their applications in asymmetric hydrogenation of beta-ketoesters.
Z Zhang et al.
The Journal of organic chemistry, 65(19), 6223-6226 (2000-09-16)
Wenjun Tang et al.
Journal of the American Chemical Society, 125(32), 9570-9571 (2003-08-09)
Hydrogenation of a series of cyclic beta-(acylamino)acrylates with tetrasubstituted olefins has been accomplished successfully with the use of Ru catalysts with chiral biaryl ligands such as C3-TunaPhos, and up to over 99% ee's have been achieved. This methodology provides an
Aiwen Lei et al.
Journal of the American Chemical Society, 126(6), 1626-1627 (2004-02-12)
A new type of alpha-phthalimide ketones was hydrogenated in excellent enantioselectivity by using a Ru-(C3-TunePhos) complex as the catalyst. Up to 10 000 turnovers have been achieved in more than 99% ee in the hydrogenation reaction. A dynamic kinetic resolution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service