695947
5,5′-Di(4-biphenylyl)-2,2′-bithiophene
97%
Synonym(s):
5,5′-[Di(1,1′-biphenyl)-4-yl]-2,2′-bithiophene
About This Item
Recommended Products
assay
97%
form
solid
mp
368-374 °C
semiconductor properties
P-type (mobility=0.04 cm2/V·s)
SMILES string
c1ccc(cc1)-c2ccc(cc2)-c3ccc(s3)-c4ccc(s4)-c5ccc(cc5)-c6ccccc6
InChI
1S/C32H22S2/c1-3-7-23(8-4-1)25-11-15-27(16-12-25)29-19-21-31(33-29)32-22-20-30(34-32)28-17-13-26(14-18-28)24-9-5-2-6-10-24/h1-22H
Inchi Key
SJNXLFCTPWBYSE-UHFFFAOYSA-N
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.
Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service