About This Item
assay
95%
form
liquid
refractive index
n20/D 1.549
density
1.465 g/mL at 25 °C
SMILES string
CCOC(=O)CC(=O)c1cc(F)cc(Br)c1
InChI
1S/C11H10BrFO3/c1-2-16-11(15)6-10(14)7-3-8(12)5-9(13)4-7/h3-5H,2,6H2,1H3
InChI key
MDZMKRYNAOMODM-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service