Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695653

Sigma-Aldrich

Salicylic acid (phenyl-13C6)

99 atom % 13C

Synonym(s):

2-Hydroxybenzoic acid (phenyl-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)13C6H4CO2H
CAS Number:
Molecular Weight:
144.08
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

mp

158-161 °C (lit.)

mass shift

M+6

SMILES string

OC(=O)[13c]1[13cH][13cH][13cH][13cH][13c]1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

YGSDEFSMJLZEOE-IDEBNGHGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service