Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695645

Sigma-Aldrich

N,N-Dimethylacetamide-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CON(CH3)2
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

99% (CP)

bp

164.5-166 °C (lit.)

mp

-20 °C (lit.)

density

0.947 g/mL at 25 °C

mass shift

M+1

SMILES string

CN(C)[13C](C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3/i4+1

InChI key

FXHOOIRPVKKKFG-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service