695572
m-Carborane-1-thiol
96%
Synonym(s):
1,7-Dicarba-closo-dodecaborane-1-yl-thiol, 1,7-Dicarbadodecaborane(12)-1-thiol, 1-Mercapto-m-carborane
About This Item
Recommended Products
assay
96%
form
solid
mp
195-209 °C
SMILES string
SC12BBBBBBBBBC1B2
InChI
1S/C2H12B10S/c13-2-1(3-2)4-6-8-10-12-11-9-7-5-2/h1,3-13H
InChI key
IKKUVNFZJOSSLF-UHFFFAOYSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service