Key Documents
695300
Sigma-Aldrich
Takasago (R)-Ru Acetate Kit I
About This Item
UNSPSC Code:
12352000
storage temp.
2-8°C
General description
Takasago (R)-Ru Acetate Kit I comprises of ruthenium-based biphenyl phosphine acetate catalysts containing either BINAP and SEGPHOS® ligands. These highly reactive and selective catalysts are useful in a variety of asymmetric reactions, mainly asymmetric hydrogenation.[1]
Legal Information
Sold in collaboration with Takasago for research purposes only.
SEGPHOS is a registered trademark of Takasago Intl. Corp.
Kit Components Also Available Separately
Product No.
Description
SDS
  • 692158(R)-Ru(OAc)2(DM-BINAP) 50 mgSDS
  • 692166(R)-Ru(OAc)2(H8-BINAP) 50 mgSDS
  • 692174(R)-Ru(OAc)2(SEGPHOS®) 50 mgSDS
  • 692182(R)-Ru(OAc)2(DM-SEGPHOS®) 50 mgSDS
  • 692220(R)-Ru(OAc)2(BINAP) 100 mgSDS
  • 692239(R)-Ru(OAc)2(T-BINAP) 100 mgSDS
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Ryoji Noyori et al.
Angewandte Chemie (International ed. in English), 40(1), 40-73 (2001-02-13)
Hydrogenation is a core technology in chemical synthesis. High rates and selectivities are attainable only by the coordination of structurally well-designed catalysts and suitable reaction conditions. The newly devised [RuCl(2)(phosphane)(2)(1,2-diamine)] complexes are excellent precatalysts for homogeneous hydrogenation of simple ketones