695130
Bis(3,5-dimethyl-4-methoxyphenyl)chlorophosphine
About This Item
form
liquid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
refractive index
n20/D 1.597
density
1.141 g/cm3 at 25 °C
functional group
phosphine
SMILES string
COc1c(C)cc(cc1C)P(Cl)c2cc(C)c(OC)c(C)c2
InChI
1S/C18H22ClO2P/c1-11-7-15(8-12(2)17(11)20-5)22(19)16-9-13(3)18(21-6)14(4)10-16/h7-10H,1-6H3
Inchi Key
JWCZKGYRAINWJA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Asymmetric organocatalytic electrophilic phosphination
- Asymmetric hydroformulation using Taddol-based chiral phosphine-phosphite ligands
- Synthesis of ferrocene-based chiral diphosphines
- Alcoholysis of phosphane-boranes
- Asymmetric hydrogenations of bifep-type biferrocenes
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service