Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

694665

Sigma-Aldrich

1-(2-Tetrahydropyranyl)-1H-pyrazole

95%

Synonym(s):

1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.503

density

1.084 g/mL at 25 °C

functional group

ether

SMILES string

C1CCC(OC1)n2cccn2

InChI

1S/C8H12N2O/c1-2-7-11-8(4-1)10-6-3-5-9-10/h3,5-6,8H,1-2,4,7H2

InChI key

IMZWSOSYNFVECD-UHFFFAOYSA-N

Application

1-THP-protected 1H-pyrazole as the valuable starter for the preparation of pyrazolylboronic ester, which is useful in the synthesis of pyrazoloisoindoles and other pyrazole derivatives[1][2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

219.9 °F

flash_point_c

104.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 4665-4665 (2006)
Mary Beth Young et al.
Journal of medicinal chemistry, 47(12), 2995-3008 (2004-05-28)
In an effort to discover potent, clinically useful thrombin inhibitors, a rapid analogue synthetic approach was used to explore the P(1) region. Various benzylamines were coupled to a pyridine/pyrazinone P(2)-P(3) template. One compound with an o-thiadiazole benzylic substitution was found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service