Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694630

Sigma-Aldrich

Lumidot CdSe/ZnS

560, core-shell type quantum dots, 5 mg/mL in toluene

Synonym(s):

QD, Quantum dot semiconductor nanocrystal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

form

dispersion

availability

not available in USA

concentration

5 mg/mL in toluene

particle size

3.4 nm

bp

110 °C

density

0.865 g/mL at 25 °C

ε (extinction coefficient)

0.97 x 105 M-1cm-1

fluorescence

λex 545 nm; λem 560 nm FWHM <40 nm, quantum yield ≥30%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Yellow emitting quantum dot nanoparticles

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

wgk_germany

WGK 3

flash_point_f

39.9 °F

flash_point_c

4.4 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service