Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694452

Sigma-Aldrich

10-(2-Tetrahydropyranyloxy)-8-decynoic acid

85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O4
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

85%

form

liquid

refractive index

n20/D 1.4843

density

1.045 g/mL at 25 °C

SMILES string

OC(=O)CCCCCCC#CCOC1CCCCO1

InChI

1S/C15H24O4/c16-14(17)10-6-4-2-1-3-5-8-12-18-15-11-7-9-13-19-15/h15H,1-4,6-7,9-13H2,(H,16,17)

InChI key

DJSRWOJODMLFNR-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service