Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694088

Sigma-Aldrich

Palladium(II) acetate-S-Phos(Pd:P 1:2), ChemDose tablets

Loading 1μmol Pd(OAc)2 per magnesium aluminometasilicate tablet

Synonym(s):

ChemDose, Palladium(II) acetate-S-Phos(Pd:P 1:2) impregnated tablets, ChemDose, Pd(OAc)2-S-Phos(Pd:P 1:2) impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

tablet

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

matrix

Magnesium aluminometasilicate base material

functional group

phosphine

storage temp.

2-8°C

SMILES string

CC(=O)O[Pd]OC(C)=O.COc1cccc(OC)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4.COc5cccc(OC)c5-c6ccccc6P(C7CCCCC7)C8CCCCC8

InChI

1S/2C26H35O2P.2C2H4O2.Pd/c2*1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;2*1-2(3)4;/h2*9-11,16-21H,3-8,12-15H2,1-2H3;2*1H3,(H,3,4);/q;;;;+2/p-2

InChI key

MHDRUZGMPXBVQJ-UHFFFAOYSA-L

Legal Information

ChemDose is a trademark of Reaxa Ltd.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service