Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694053

Sigma-Aldrich

Tris[dibenzylideneacetone]dipalladium(0)-S-Phos (Pd:P 1:2), ChemDose tablets

Loading 0.5μmol Pd2(dba)3 per magnesium aluminometasilicate tablet (1μmol Pd per tablet)

Synonym(s):

ChemDose, Pd2(dba)3-S-Phos (Pd:P 1:2) impregnated tablets, ChemDose, Tris[dibenzylideneacetone]dipalladium(0)-S-Phos (Pd:P 1:2) impregnated tablets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

form

tablet

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

matrix

Magnesium aluminometasilicate base material

functional group

phosphine

storage temp.

2-8°C

SMILES string

[Pd][Pd].O=C(/C=C/c1ccccc1)\C=C\c2ccccc2.O=C(/C=C/c3ccccc3)\C=C\c4ccccc4.O=C(/C=C/c5ccccc5)\C=C\c6ccccc6.COc7cccc(OC)c7-c8ccccc8P(C9CCCCC9)C%10CCCCC%10.COc%11cccc(OC)c%11-c%12ccccc%12P(C%13CCCCC%13)C%14CCCCC%14.COc%15cccc(OC)c%15-c%16ccccc%16P(C%17CCCCC%17)C%18CCCCC%18.COc%19cccc(OC)c%19-c%20ccccc%20P(C%21CCCCC%21)C%22CCCCC%22

InChI

1S/4C26H35O2P.3C17H14O.2Pd/c4*1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;;/h4*9-11,16-21H,3-8,12-15H2,1-2H3;3*1-14H;;/b;;;;3*13-11+,14-12+;;

InChI key

HGHLKXZFMABJGY-NLFLYQHWSA-N

Legal Information

ChemDose is a trademark of Reaxa Ltd.

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service