Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694029

Sigma-Aldrich

1-(Methyldithiocarbonyl)imidazole

97%

Synonym(s):

1H-Imidazole-1-carbodithioic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2S2
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

reaction type: Carbonylations
reaction type: Coupling Reactions

refractive index

n20/D 1.6741

mp

29-33 °C

density

1.294 at 25 °C

SMILES string

CSC(=S)n1ccnc1

InChI

1S/C5H6N2S2/c1-9-5(8)7-3-2-6-4-7/h2-4H,1H3

InChI key

ZSKBYJIWBCCFMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service