storage temp.
−20°C
Legal Information
Kit Components Also Available Separately
- 665290(S,S,S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a’]dinaphthalen-4-yl)bis(1-phenylethyl)amine, 97% 100 mgSDS
- 665347(S,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)-(1-phenylethyl)amine, 96% 100 mgSDS
- 665355(S)-(+)-Benzyl(3,5-dioxa-4-phospha-cyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)methylamine, 97% 100 mgSDS
- 665363(S,R,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine, 95% (HPLC) 100 mgSDS
- 665460(3aR,8aR)-(−)-(2,2-Dimethyl-4,4,8,8-tetraphenyl-tetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)dimethylamine, 96% 100 mgSDS
- (S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)piperidine, 97% 100 mg
- (S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4-a′]dinaphthalen-4-yl)morpholine, 97% 100 mg
- 668192(S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a;3,4- a′]dinaphthalen-4-yl)dimethylamine, 97% 250 mgSDS
- 668206(R)-(−)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)dimethylamine, 97% 250 mgSDS
- 683248(11bR)-2,6-Bis(diphenylphosphino)-N,N-dimethyldinaphtho[2,1-d:1′,2′-f]-1,3,2-dioxaphosphepin-4-amine 100 mgSDS
- 685569(11bS)-N,N-dimethyl-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine 100 mgSDS
Storage Class
13 - Non Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service