Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693731

Sigma-Aldrich

Samarium

foil, thickness 1 mm, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace rare earth metals basis

form

foil

reaction suitability

reagent type: catalyst
core: samarium

resistivity

91.4 μΩ-cm, 0°C

thickness

1 mm

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

Inchi Key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Quantity

1 ea = 25 x 25 mm

Pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Water-react 3

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Lu et al.
Optics express, 21(2), 1997-2002 (2013-02-08)
We demonstrate an all-fiber passively Q-switched Yb-doped laser using a piece of Sm-doped fiber as a saturable absorber. The laser was pumped by two 25W, 975 nm fiber coupled diodes and Q-switching was initiated when the ASE generated in the
M Saif et al.
Chemosphere, 90(2), 840-847 (2012-11-06)
Highly active samarium doped zinc oxide self-cleaning and biocidal surfaces (x mol% Sm(3+)/ZnO where x=0, 1, 2 and 4 mol%) with crystalline porous structures were synthesized by hydrothermal method. Sm(3+)/ZnO thin films were characterized by X-ray diffraction (XRD), transmission electron
Hua Yang et al.
Inorganic chemistry, 52(3), 1275-1284 (2013-01-25)
Sm@C(2v)(3)-C(80) has been separated from the carbon soot produced by electrical arc vaporization of graphite rods doped with Sm(2)O(3) and purified. Its structure has been determined by single crystal X-ray diffraction using cocrystals obtained from either Ni(II)(octaethylporphyrin) (Ni(II)(OEP)) to form
M Majumder et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(2), 025701-025701 (2012-12-05)
We report resistivity, magnetization and (19)F NMR results in a polycrystalline sample of SmFeAsO(0.86)F(0.14). The resistivity and magnetization data show a sharp drop at 48 K indicating a superconducting transition. The nuclear spin-lattice rate (1/T(1)) and spin-spin relaxation rate (1/T(2))
I Pal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 101, 74-81 (2012-10-27)
The glasses with compositions 20ZnO·(79.5-x)Bi2O3·xSiO2·0.5Sm2O3 (10≤x≤50, mol%) have been synthesized using normal melt-quench technique. Optical absorption and fluorescence spectra of the glasses were recorded at ambient temperature. Judd-Ofelt (J-O) theory has been successfully applied to characterize the absorption and luminescence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service