Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693642

Sigma-Aldrich

Yttrium

foil, thickness 1 mm, 99.8% trace rare earth metals basis

Synonym(s):

Yttrium element, Yttrium-89

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.8% trace rare earth metals basis

form

foil

reaction suitability

reagent type: catalyst
core: yttrium

resistivity

57 μΩ-cm, 20°C

thickness

1 mm

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Quantity

1 ea = 25 x 25 mm

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yttrium-catalyzed addition of benzylic C-H bonds of alkyl pyridines to olefins.
Bing-Tao Guan et al.
Angewandte Chemie (International ed. in English), 52(16), 4418-4421 (2013-03-21)
Masahiko Ooe et al.
Aesthetic plastic surgery, 37(2), 424-433 (2013-02-12)
Skin wrinkles are one of the most cosmetically concerning signs of aging for women, and improvements in the visual effect of wrinkles become a matter of concern that has an impact on the quality of life. Although various wrinkle treatments
José Renato Queiroz et al.
The journal of adhesive dentistry, 15(2), 151-159 (2013-03-28)
To compare the effect of silica (Si)-based nano-coating deposited by reactive magnetron sputtering (RMP) with that of conventional surface conditioning using metal/zirconia primer alone or after air-particle abrasion on the adhesion of resin cements to zirconia ceramic. Two hundred forty
S Blanco-Canosa et al.
Physical review letters, 110(18), 187001-187001 (2013-05-21)
We use resonant x-ray scattering to determine the momentum-dependent charge correlations in YBa(2)Cu(3) O(6.55) samples with highly ordered chain arrays of oxygen acceptors (ortho-II structure). The results reveal nearly critical, biaxial charge density wave (CDW) correlations at in-plane wave vectors
Jianyuan Zhang et al.
Nature chemistry, 5(10), 880-885 (2013-09-24)
Although fullerenes were discovered nearly three decades ago, the mechanism of their formation remains a mystery. Many versions of the classic 'bottom-up' formation mechanism have been advanced, starting with C2 units that build up to form chains and rings of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service