Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693227

Sigma-Aldrich

RuCl2[(S)-(DM-BINAP)][(S)-DAIPEN]

Synonym(s):

Dichloro[(S)-(−)-2,2′-bis[di(3,5-xylyl)phosphino]-1,1′-binaphthyl][(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C71H74Cl2N2O2P2Ru
CAS Number:
Molecular Weight:
1221.28
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2([NH2][Ru]3(Cl)(Cl)([NH2][C@H]2C(C)C)[PH](c4cc(C)cc(C)c4)(c5cc(C)cc(C)c5)c6ccc7ccccc7c6-c8c(ccc9ccccc89)[PH]3(c%10cc(C)cc(C)c%10)c%11cc(C)cc(C)c%11)c%12ccc(OC)cc%12

InChI

1S/C52H48P2.C19H26N2O2.2ClH.Ru/c1-33-21-34(2)26-43(25-33)53(44-27-35(3)22-36(4)28-44)49-19-17-41-13-9-11-15-47(41)51(49)52-48-16-12-10-14-42(48)18-20-50(52)54(45-29-37(5)23-38(6)30-45)46-31-39(7)24-40(8)32-46;1-13(2)18(20)19(21,14-5-9-16(22-3)10-6-14)15-7-11-17(23-4)12-8-15;;;/h9-32H,1-8H3;5-13,18H,20-21H2,1-4H3;2*1H;/t;18-;;;/m.0.../s1

InChI key

VMSNEARTLSFOHB-OGLOXHGMSA-N

Application

Catalyst for:
  • Preparation of cyclometalated ruthenium 1,1-dianisyl-2-isopropyl-1,2-ethylenediamine complexes via asymmetric hydrogenation of ketones
  • Enantioselective hydrogenation of an α-alkoxy substituted ketone with chiral ruthenium (phosphinoferrocenyl)oxazoline complexes
  • Nonclassical asymmeteric hydrogen transfer between alcohols and carbonyl compounds
  • Asymmetric hydrogenation of amino and heteroaromatic ketones

  • Pharmacologically active substituted oxazolidinone used as NPC1L1 ligand for inhibition of cholesterol absorption

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 2041996, 2731377, 2935453 JP Appllication No. 19970359654

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service