Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693146

Sigma-Aldrich

(S)-[(RuCl(T-BINAP))2(μ-Cl)3[NH2Me2]

Synonym(s):

Dimethylammonium dichlorotri(μ-chloro)bis[(S)-(−)-2,2′-bis(di-p-tolylphosphino)-1,1′-binaphthyl]diruthenate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH2+[C96H80Cl5P4Ru2]-
CAS Number:
Molecular Weight:
1783.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

storage temp.

2-8°C

InChI

1S/2C48H40P2.C2H7N.2ClH.3Cl.2Ru/c2*1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42;1-3-2;;;;;;;/h2*5-32H,1-4H3;3H,1-2H3;2*1H;;;;;/q;;;;;3*+1;2*-1/p-1

InChI key

QGBBTZBFEYSGDQ-UHFFFAOYSA-M

Legal Information

Sold in collaboration with Takasago for research purposes only.

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service