Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692956

Sigma-Aldrich

(S)-RuCl[(p-cymene)(SEGPHOS®)]Cl

Synonym(s):

Chloro[(S)-(−)-5,5′-bis(diphenylphosphino)-4,4′-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C48H42ClO4P2Ru]+Cl
CAS Number:
Molecular Weight:
916.77
UNSPSC Code:
12352101
PubChem Substance ID:

form

powder

storage temp.

2-8°C

SMILES string

[Cl-].Cl[Ru+].C[C]1[CH][CH][C]([CH][CH]1)C(C)C.C2Oc3ccc(P(c4ccccc4)c5ccccc5)c(c3O2)-c6c7OCOc7ccc6P(c8ccccc8)c9ccccc9

InChI

1S/C38H28O4P2.C10H14.2ClH.Ru/c1-5-13-27(14-6-1)43(28-15-7-2-8-16-28)33-23-21-31-37(41-25-39-31)35(33)36-34(24-22-32-38(36)42-26-40-32)44(29-17-9-3-10-18-29)30-19-11-4-12-20-30;1-8(2)10-6-4-9(3)5-7-10;;;/h1-24H,25-26H2;4-8H,1-3H3;2*1H;/q;;;;+2/p-2

InChI key

HZLSGDCLBWZKRB-UHFFFAOYSA-L

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service