692913
Bis[(dicyclohexyl)(4-dimethylaminophenyl)phosphine] palladium(II) chloride
96%
Synonym(s):
(A-caPhos)2 PdCl2
About This Item
Recommended Products
assay
96%
form
powder
reaction suitability
core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings
functional group
phosphine
SMILES string
CN(C)c1ccc(cc1)[PH](C2CCCCC2)(C3CCCCC3)[Pd](Cl)(Cl)[PH](C4CCCCC4)(C5CCCCC5)c6ccc(cc6)N(C)C
InChI
1S/2C20H32NP.2ClH.Pd/c2*1-21(2)17-13-15-20(16-14-17)22(18-9-5-3-6-10-18)19-11-7-4-8-12-19;;;/h2*13-16,18-19H,3-12H2,1-2H3;2*1H;
InChI key
ZVLBDYRPWSIJCO-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Ligand used to prepare a palladium dichloride catalyst on treatment with PdCl2(COD). The catalyst effectively cross-couples aryl boronic acids with heteroaryl chlorides.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service