Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692867

Sigma-Aldrich

Aluminum nickel cobalt

alloy 11, disc, size 13 mm × 6 mm

Synonym(s):

AlNiCo

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
NACRES:
NA.23

form

disc

composition

Aluminum, 7.5-8.7 wt. %
Cobalt, 23.5-25 wt. %
Copper, 2-3.5 wt. %
Iron, 45.3-52.85 wt. %
Nickel, 12.5-15.5 wt. %
Silicon, 1.5 wt. %
Titanium, 0.15-0.5 wt. %

size

13 mm × 6 mm

SMILES string

[Al].[Co].[Ni]

InChI

1S/Al.Co.Ni

InChI key

VKLQGCFJXKUNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Magnetic parameters/properties:
• Maximum Energy Product (BH): 84 kJ/m3 (10.6 MGsOe)
• Residual Induction (Br): 1.12 T (11.2 kGs)
• Coercive Force (Hc): 109 kA/m (1.37 kOe)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service