Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

692611

Sigma-Aldrich

4-[(Phenylimino)methyl]phenylboronic acid pinacol ester

97%

Synonym(s):

N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene]benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22BNO2
Molecular Weight:
307.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

powder

mp

96-101 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(cc2)\C=N\c3ccccc3

InChI

1S/C19H22BNO2/c1-18(2)19(3,4)23-20(22-18)16-12-10-15(11-13-16)14-21-17-8-6-5-7-9-17/h5-14H,1-4H3/b21-14+

InChI key

QOACLCIBJOVFJC-KGENOOAVSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service