Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691518

Sigma-Aldrich

2-Piperidineacetic acid hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14ClNO2
CAS Number:
Molecular Weight:
179.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

183-186 °C (lit.)

functional group

carboxylic acid

SMILES string

Cl.OC(=O)CC1CCCCN1

InChI

1S/C7H13NO2.ClH/c9-7(10)5-6-3-1-2-4-8-6;/h6,8H,1-5H2,(H,9,10);1H

Inchi Key

DJEVRGSQHJHEKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Enantiopure piperidine based β-amino esters via chemoenzymatic approach
  • Esterification

Pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service