Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691119

Sigma-Aldrich

3-Ethoxy-acryloyl chloride

≥85% (H-NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7ClO2
CAS Number:
Molecular Weight:
134.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥85% (H-NMR)

form

liquid

functional group

acyl chloride
ether

storage temp.

2-8°C

SMILES string

CCO\C=C\C(Cl)=O

InChI

1S/C5H7ClO2/c1-2-8-4-3-5(6)7/h3-4H,2H2,1H3/b4-3+

InChI key

SFMFACMIOWQIPR-ONEGZZNKSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

77.0 °F

flash_point_c

25 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service