Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690988

Sigma-Aldrich

1,5-Cyclooctadiene{[dibenzyl((4R,5R)-5-methyl-2-phenyl-4,5-dihydro-4-oxazolyl)methyl]diphenylphosphinite κN:κP}iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate

97%

Synonym(s):

Ubaphox, [((4R,5R)-Ph2-Ubaphox)Ir(COD)]BARF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H58BF24IrNO2P
CAS Number:
Molecular Weight:
1719.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

form

lumps

optical purity

ee: ≥99%

storage condition

under inert gas

storage temp.

2-8°C

SMILES string

[Ir+].C1CC=CCCC=C1.C[C@H]2OC(=N[C@H]2C(Cc3ccccc3)(Cc4ccccc4)OP(c5ccccc5)c6ccccc6)c7ccccc7.FC(F)(F)c8cc(cc(c8)C(F)(F)F)[B-](c9cc(cc(c9)C(F)(F)F)C(F)(F)F)(c%10cc(cc(c%10)C(F)(F)F)C(F)(F)F)c%11cc(cc(c%11)C(F)(F)F)C(F)(F)F

InChI

1S/C37H34NO2P.C32H12BF24.C8H12.Ir/c1-29-35(38-36(39-29)32-21-11-4-12-22-32)37(27-30-17-7-2-8-18-30,28-31-19-9-3-10-20-31)40-41(33-23-13-5-14-24-33)34-25-15-6-16-26-34;34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;1-2-4-6-8-7-5-3-1;/h2-26,29,35H,27-28H2,1H3;1-12H;1-2,7-8H,3-6H2;/q;-1;;+1/b;;2-1-,8-7-;/t29-,35-;;;/m1.../s1

InChI key

RWJSSAILCUYYLD-XCHXSORBSA-N

General description

sold in collaboration with Solvias AG

Application

Catalyst for enantioselective hydrogenation of alkenes

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Threonine-derived phosphinite-oxazoline ligands for the Ir-catalyzed enantioselective hydrogenation.
Menges, F. and Pfaltz, A.
Advanced Synthesis & Catalysis, 344, 40-44 (2002)
A New Class of Modular Phosphinite-Oxazoline Ligands: Ir-Catalyzed Enantioselective Hydrogenation of Alkenes We thank Dr. Martin Studer and Dr. Benoît Pugin (Solvias AG, Basel) for fruitful discussions, and Prof. Kevin Burgess (Texas A&M University) for preprints of related unpublished work. Financial support by the Swiss National Science Foundation is gratefully acknowledged.
Jörg Blankenstein et al.
Angewandte Chemie (International ed. in English), 40(23), 4445-4447 (2002-10-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service