Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690937

Sigma-Aldrich

1,3-Dimethoxyimidazolium bis(trifluoromethyl­sulfonyl)imide

98%, complies for IR spectroscopy

Synonym(s):

(OMe)2Im-NTf2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9F6N3O6S2
CAS Number:
Molecular Weight:
409.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98% (13C-NMR)
≥98% (19F-NMR)
≥98% (31P-NMR)
98%

form

liquid

impurities

0.8% Pf6 (19F-NMR)

refractive index

n20/D 1.4249

suitability

complies for IR spectroscopy

SMILES string

COn1cc[n+](OC)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C5H9N2O2.C2F6NO4S2/c1-8-6-3-4-7(5-6)9-2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h3-5H,1-2H3;/q+1;-1

InChI key

PPGPWZWXCSNVNK-UHFFFAOYSA-N

Application

Ionic liquid and NHC precatalyst[1]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service