Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690686

Sigma-Aldrich

N-Benzylidene-p-toluidine

≥96.0%

Synonym(s):

4-Methyl-N-(phenylmethylene)benzenamine, N-Benzylidene-4-methylaniline, Benzylidene-p-tolyl-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥96.0% (GC)
≥96.0%
96.0-104.0% (NT)

form

solid

SMILES string

CC1=CC=C(/N=C/C2=CC=CC=C2)C=C1

InChI

1S/C14H13N/c1-12-7-9-14(10-8-12)15-11-13-5-3-2-4-6-13/h2-11H,1H3/b15-11+

InChI key

MSFVFFZPHJPOHP-RVDMUPIBSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic N-oxidation of diarylimines: a new metabolic reaction of potential toxicological significance.
J W Gorrod et al.
Drug metabolism reviews, 26(1-2), 241-252 (1994-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service