Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69052

Sigma-Aldrich

Vanadyl acetylacetonate bound to polyaniline

extent of labeling: ~1.0 mmol/g loading

Synonym(s):

Vanadium(IV)-oxy acetylacetonate bound to polyaniline

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

crystals

Quality Level

reaction suitability

core: vanadium
reagent type: catalyst

extent of labeling

~1.0 mmol/g loading

matrix

polyaniline

particle size

200-400 mesh

Application

Catalyst for the oxidation of alcohols to aldehydes and ketones with molecular oxygen[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reddy, S.R., et al.
Tetrahedron Letters, 45, 3561-3564 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service