Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690333

Sigma-Aldrich

(S)-N-(α-Methylbenzyl)hydroxylamine oxalate salt

≥96.0%

Synonym(s):

R)-N-Hydroxy-α-methyl-benzenemethanamine ethanedioate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO· C2H2O4
CAS Number:
Molecular Weight:
227.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

≥96.0% (HPLC)
≥96.0%

optical purity

enantiomeric excess: ≥99.0% (HPLC)

SMILES string

OC(=O)C(O)=O.C[C@H](NO)c1ccccc1

InChI

1S/C8H11NO.C2H2O4/c1-7(9-10)8-5-3-2-4-6-8;3-1(4)2(5)6/h2-7,9-10H,1H3;(H,3,4)(H,5,6)/t7-;/m0./s1

InChI key

GUXMGLCOBSBIKH-FJXQXJEOSA-N

Pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service