Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69006

Sigma-Aldrich

1,4-Diisocyanatobutane

technical, ≥95.0% (GC)

Synonym(s):

Tetramethylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)4NCO
CAS Number:
Molecular Weight:
140.14
Beilstein/REAXYS Number:
1756894
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.453

bp

102-104 °C/14 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NCCCCN=C=O

InChI

1S/C6H8N2O2/c9-5-7-3-1-2-4-8-6-10/h1-4H2

InChI key

OVBFMUAFNIIQAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service