689688
(1R,2S)-trans-2-(Fmoc-aminomethyl)cyclohexanol
97%
About This Item
assay
≥96.5% (GC)
97%
form
powder
optical purity
enantiomeric excess: ≥97.5%
functional group
Fmoc
storage temp.
2-8°C
SMILES string
O[C@@H]1CCCC[C@H]1CNC(=O)OCC2c3ccccc3-c4ccccc24
InChI
1S/C22H25NO3/c24-21-12-6-1-7-15(21)13-23-22(25)26-14-20-18-10-4-2-8-16(18)17-9-3-5-11-19(17)20/h2-5,8-11,15,20-21,24H,1,6-7,12-14H2,(H,23,25)/t15-,21+/m0/s1
InChI key
DLDUSAROVXDNNQ-YCRPNKLZSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service